missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-fluoro-2-phosphonoacetate, 96%

Product Code. 11354825
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354825 1 g 1g
11364825 5 g 5g
11374825 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354825 Supplier Thermo Scientific Alfa Aesar Supplier No. L17913.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2356-16-3 | C8H16FO5P | 242.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2356-16-3
Molecular Formula C8H16FO5P
Molecular Weight (g/mol) 242.183
MDL Number MFCD00134400
InChI Key FVPISMANESAJQZ-UHFFFAOYSA-N
Synonym triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester
PubChem CID 2734865
IUPAC Name ethyl 2-diethoxyphosphoryl-2-fluoroacetate
SMILES CCOC(=O)C(F)P(=O)(OCC)OCC

Specifications

Density 1.194
Boiling Point 75°C (0.01 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.425
Quantity 1 g
Beilstein 1912161
Formula Weight 242.19
Percent Purity 96%
Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.