missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonopentanoate, 98%

Product Code. 11317278
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317278 10 g 10g
11307278 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317278 Supplier Thermo Scientific Alfa Aesar Supplier No. 030413.09
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35051-49-1 | C11H23O5P | 266.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35051-49-1
Molecular Formula C11H23O5P
Molecular Weight (g/mol) 266.274
MDL Number MFCD00015163
InChI Key BUPVIVDUPRDDFI-UHFFFAOYSA-N
Synonym triethyl 2-phosphonopentanoate, ethyl 2-diethoxyphosphoryl pentanoate, pentanoic acid, 2-diethoxyphosphinyl-, ethyl ester, valeric acid, 2-diethylphosphono-, ethyl ester, alpha-carbethoxybutylphosphonic acid, diethyl ester, triethyl 2-phosphonovalerate, phosphonic acid, alpha-carbethoxybutyl-, diethyl ester, ethyl 2-diethoxyphosphoryl pentanoate #, diethyl 1-ethoxycarbonyl butylphosphonate
PubChem CID 98244
IUPAC Name ethyl 2-diethoxyphosphorylpentanoate
SMILES CCCC(C(=O)OCC)P(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 98°C (2 mmHg)
Linear Formula (C2H5O)2P(O)[CH(C3H7)CO2C2H5]
Quantity 10 g
Formula Weight 266.27
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl 2-phosphonopentanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.