missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 3-phosphonopropionate, 98%

Product Code. 11378285
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378285 10 g 10g
11388285 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378285 Supplier Thermo Scientific Alfa Aesar Supplier No. L10190.09
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3699-67-0 | C9H19O5P | 238.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3699-67-0
Molecular Formula C9H19O5P
Molecular Weight (g/mol) 238.22
MDL Number MFCD00015167
InChI Key JMJWCUOIOKBVNQ-UHFFFAOYSA-N
Synonym ethyl 3-diethoxyphosphoryl propanoate, triethyl 3-phosphonopropionate, 3-diethylphosphonopropionic acid ethyl ester, triethyl 3-phosphonopropanoate, ethyl 3-diethoxyphosphinyl propionate, ethyl 3-diethoxycarbonyl propanoate, propanoic acid, 3-diethoxyphosphinyl-, ethyl ester, ethyl 3-diethylphosphonopropionate, ethyl 3-diethylphosphono-propionate, ethyl 3-diethylphosphono propanoate
PubChem CID 281204
IUPAC Name ethyl 3-diethoxyphosphorylpropanoate
SMILES CCOC(=O)CCP(=O)(OCC)OCC

Specifications

Density 1.094
Boiling Point 127°C to 129°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.433
Quantity 10 g
Beilstein 1790966
Formula Weight 238.22
Percent Purity 98%
Chemical Name or Material Triethyl 3-phosphonopropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.