missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 4-phosphonobutyrate, 97%

Product Code. 11378455
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378455 10 g 10g
11368455 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378455 Supplier Thermo Scientific Alfa Aesar Supplier No. L10488.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2327-69-7 | C10H21O5P | 252.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2327-69-7
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00015168
InChI Key FYESTJSRASJZOY-UHFFFAOYSA-N
PubChem CID 281201
IUPAC Name ethyl 4-diethoxyphosphorylbutanoate
SMILES CCOC(=O)CCCP(=O)(OCC)OCC

Specifications

Boiling Point 117°C to 119°C (0.4 mmHg)
Refractive Index 1.437
Quantity 10 g
Beilstein 1791802
Synonym triethyl 4-phosphonobutyrate, ethyl 4-diethoxyphosphoryl butanoate, triethyl4-phosphonobutyrate, butanoic acid, 4-diethoxyphosphinyl-, ethyl ester, triethyl-4-phosphono butyrate, butyric acid, 4-diethylphosphono-, ethyl ester, triethyl 4-phosphonobutanoate, 4-phosphonobutanoic acid triethyl ester, 4-diethoxyphosphinyl butyric acid ethyl ester, butanoic acid,4-diethoxyphosphinyl-,ethyl ester
Formula Weight 252.25
Percent Purity 97%
Chemical Name or Material Triethyl 4-phosphonobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.