missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 13102174
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
122-51-0
C7H16O3
148.2
GKASDNZWUGIAMG-UHFFFAOYSA-N
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
31214
diethoxymethoxyethane
CCOC(OCC)OCC
This item is not returnable. View return policy
122-51-0
C7H16O3
148.2
GKASDNZWUGIAMG-UHFFFAOYSA-N
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
31214
diethoxymethoxyethane
CCOC(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -61.0°C
Density 0.8900g/mL
Boiling Point 146.0°C
Flash Point 35°C
Assay Percent Range 97.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH(OC2H5)3
Refractive Index 1.3900 to 1.3920
Quantity 100 mL
Merck Index 15,698
Specific Gravity 0.89
Solubility Information Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform,
Formula Weight 148.2
Percent Purity 98%
Chemical Name or Material Triethyl orthoformate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl orthoformate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'