missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl orthoformate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Specifications
Melting Point | -61.0°C |
Color | Colorless |
Density | 0.8900g/mL |
Boiling Point | 146.0°C |
Flash Point | 35°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH(OC2H5)3 |
Refractive Index | 1.3900 to 1.3920 |
Show More |
Safety and Handling
Product Identifier
- Triethyl orthoformate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : Triethyl orthoformate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'