missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphate, 99%

Product Code. 10143190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143190 2.5 kg Glass bottle 2.5kg
10522731 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143190 Supplier Thermo Scientific Acros Supplier No. 139680025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Melting Point -56°C
Density 1.0700g/mL
Boiling Point 215°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)3P(O)
Refractive Index 1.4050 to 1.407
Quantity 2.5 kg
Beilstein 01, 332
Fieser 01,1212; 16,3
Merck Index 15, 9838
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols,ethers,ketones,esters,,aliphatic,aromatic and chlorinated hydrocarbons,soluble in most other organic solvents
Formula Weight 182.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethyl phosphate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.