missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphonoacetate, 97%

Product Code. 11915801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915801 2.5 kg Glass bottle 2.5kg
10467340 100 g Glass bottle 100g
10684612 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11915801 Supplier Thermo Scientific Acros Supplier No. 139700025
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 867-13-0 | C8H17O5P | 224.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-13-0
Molecular Formula C8H17O5P
Molecular Weight (g/mol) 224.19
InChI Key GGUBFICZYGKNTD-UHFFFAOYSA-N
Synonym triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
PubChem CID 13345
IUPAC Name ethyl 2-diethoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OCC)OCC

Specifications

Acidity 2mg KOH/g max.
Melting Point -24.0°C
Density 1.1300g/mL
Boiling Point 260.0°C to 262.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
Refractive Index 1.4300 to 1.4320
Quantity 2.5 kg
Beilstein 04,I,573
Fieser 01,1216; 06,612; 12,527; 13,255
Specific Gravity 1.13
Solubility Information Solubility in water: insoluble
Formula Weight 224.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl phosphonoacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.