missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphonoacetate, 98+%

Product Code. 11458086
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458086 2500 g 2500g
11438086 100 g 100g
11448086 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458086 Supplier Thermo Scientific Alfa Aesar Supplier No. A14120.0E
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 867-13-0 | C8H17O5P | 224.193 g/mol

Triethyl phosphonoacetate serves as a reactant used in Horner-Wadsworth-Emmons reactions, Tsuji-Trost type reactions, Intramolecular Heck-type cyclization and isomerizations and Intramolecular aryne-ene reactions. In Horner-Wadsworth-Emmons reaction, it is utilized as a reagent to prepare chiral 2-methylcyclopropanecarboxylic acid from (S)-propylene oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphonoacetate serves as a reactant used in Horner-Wadsworth-Emmons reactions, Tsuji-Trost type reactions, Intramolecular Heck-type cyclization and isomerizations and Intramolecular aryne-ene reactions. In Horner-Wadsworth-Emmons reaction, it is utilized as a reagent to prepare chiral 2-methylcyclopropanecarboxylic acid from (S)-propylene oxide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-13-0
Molecular Formula C8H17O5P
Molecular Weight (g/mol) 224.193
MDL Number MFCD00009177
InChI Key GGUBFICZYGKNTD-UHFFFAOYSA-N
Synonym triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
PubChem CID 13345
IUPAC Name ethyl 2-diethoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OCC)OCC

Specifications

Melting Point -24°C
Density 1.125
Boiling Point 260°C to 262°C
Flash Point 165°C (329°F)
Refractive Index 1.431
Quantity 2500 g
Beilstein 1343714
Solubility Information Slightly miscible with water.
Formula Weight 224.19
Percent Purity ≥98%
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.