missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Phosphonoacetate, TRC

Product Code. 30454117
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
Unit Size:
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454117 25 g -
30453310 500 g 500g
30449593 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454117 Supplier Toronto Research Chemicals Supplier No. TRCT77700025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-13-0
Molecular Formula C8 H17 O5 P
Molecular Weight (g/mol) 224.19
Synonym Phosphonoacetic Acid-Triethyl Ester, Acetic acid, (diethoxyphosphinyl)-, ethyl ester (9CI), Acetic acid, phosphono-, triethyl ester (6CI,7CI,8CI), (Diethoxyphosphinyl)acetic acid ethyl ester, (Diethoxyphosphoryl)acetic acid ethyl ester, (Ethoxycarbonylmethyl)diethoxyphosphine oxide, 2-Phosphonoacetic acid triethyl ester, Carbethoxymethyldiethyl phosphonate, Diethyl 2-ethoxy-2-oxoethylphosphonate, Diethyl carbethoxymethylphosphonate, Diethyl ethoxycarbonylmethanephosphonate, Diethyl phosphonoacetic acid ethyl ester, Ethyl (diethoxyphosphinyl)acetate, Ethyl (diethylphosphono)acetate, Ethyl 2-(diethoxylphosphoryl)acetate, Ethyl 2-(diethoxyphosphinyl)acetate, Ethyl 2-(diethoxyphosphoryl)acetate, Ethyl 2-(diethylphosphono)acetate, Ethyl diethoxyphosphoryl acetate, Ethyl α-diethylphosphonoacetate, JC 224, NSC 13898, NSC 16128, Phosphonoacetic acid triethyl ester, Triethyl 2-phosphonoacetate, Triethyl phosphonacetate, Triethyl phosphonoacetate
IUPAC Name ethyl 2-diethoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OCC)OCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H17O5P/c1-4-11-8(9)7-14(10,12-5-2)13-6-3/h4-7H2,1-3H3
Formula Weight 224.0814
Chemical Name or Material Triethyl Phosphonoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.