missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99%, pure

Product Code. 10046651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10046651 5 mL Glass bottle 5mL
10651193 100 mL Glass bottle 100mL
10265820 1 L EcoSafPak™ Glass Bottle 1 L
10626362 2.5 L Glass bottle 2.5 L
11905181 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10046651 Supplier Thermo Scientific Acros Supplier No. 157910050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-44-8 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Refractive Index 1.3990 to 1.4010
Quantity 5 mL
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Merck Index 15, 9831
Specific Gravity 0.728
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.19
Viscosity 0.36 mPa.s (20°C)
Formula Weight 101.19
Vapor Pressure 69mbar at 20°C
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Triethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.