missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99+%

Código de producto. 11341779
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg
Tamaño de la unidad:
1kg
250g
3kg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11341779

Marca: Thermo Scientific Alfa Aesar 012391.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 121-44-8 | C6H15N | 101.193 g/mol

Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Especificaciones

Melting Point -115°C
Color Colorless
Density 0.726 g/mL
Boiling Point 89°C to 90°C
Flash Point -11°C (12°F)
Assay Percent Range >99%
Odor Ammonia-like
Linear Formula (CH3CH2)3N
Refractive Index 1.4005
Quantity 1 kg
UN Number UN1296
Beilstein 1843166
Merck Index 14,9666
Solubility Information Miscible with water,ether and ethanol.
Formula Weight 101.19
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Triethylamine
Mostrar más Mostrar menos

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado