missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, Eluent Additive For Laboratory Use and LC-MS, Chem Lab

Product Code. 15949336
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15949336 50 mL 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949336 Supplier Chem-Lab Supplier No. CL00.2049.0050
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifikationer

CAS 121-44-8
Melting Point -115°C
Color Colorless
Density 0.73 g/mL
Boiling Point 90°C
Flash Point -11 °C
Assay Percent Range > 99.7%
Packaging Glass bottles
Molecular Formula C6H15N
Quantity 50 mL
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES CCN(CC)CC
IUPAC Name triethylamine
Molecular Weight (g/mol) 101.19
Physical Form Liquid
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Triethylamine, eluent additive for LC-MS
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contain: Triethylamine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.