missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, Fisher BioReagents

Product Code. 10649933
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10649933 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10649933 Supplier Fisher BioReagents Supplier No. BP616500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Triethylamine is used as a solvent in automated synthesis protocols.
TRUSTED_SUSTAINABILITY

Specifications

CAS 121-44-8
Melting Point -115°C
Color Colorless
Density 0.728g/cm³
pH 12.4
Boiling Point 90°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Refractive Index 1.397 to 1.403 (at 25°C)
Quantity 500 mL
Merck Index 15, 9831
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES CCN(CC)CC
IUPAC Name N,N-diethylethanamine
Molecular Weight (g/mol) 101.193
PubChem CID 8471
ChEBI CHEBI:35026
Formula Weight 101.19
Vapor Pressure 69mbar at 20°C
Percent Purity ≥99%
Grade Peptide Synthesis
Physical Form Liquid
Chemical Name or Material Triethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.