missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, for HPLC, ≥99.5%, Honeywell Fluka™

Product Code. 16683944 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16683944 100 mL 100mL
16693944 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16683944 Supplier Honeywell Fluka Supplier No. 17924100ml
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym N,N-Diethylethanamine
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

CAS Max % 1
Melting Point -115°C
Color Colorless to Yellow
Density 0.73 g/cm3 (20°C)
pH 12.7
Boiling Point 90°C
Flash Point -11°C
Linear Formula (C2H5)3N
Quantity 100 mL
Beilstein 605283
Grade HPLC Buffer Salt
Chemical Name or Material Triethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.