missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine hydrobromide, 98%

Product Code. 11489046
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489046 100 g 100g
11499046 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489046 Supplier Thermo Scientific Alfa Aesar Supplier No. A17926.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 636-70-4 | C6H16BrN | 182.11 g/mol

Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.

Solubility
Soluble in water, alcohol and chloroform.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-70-4
Molecular Formula C6H16BrN
Molecular Weight (g/mol) 182.11
MDL Number MFCD00054287
InChI Key NRTLTGGGUQIRRT-UHFFFAOYSA-N
Synonym triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
PubChem CID 2734074
IUPAC Name N,N-diethylethanamine;hydrobromide
SMILES [Br-].CC[NH+](CC)CC

Specifications

Melting Point ∼250°C (decomposition)
Odor Amine-like
Linear Formula (CH3CH2)3N·HBr
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol and chloroform.
Formula Weight 182.1
Percent Purity 98%
Chemical Name or Material Triethylamine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.