missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine hydrobromide, 99%

Product Code. 10517021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10517021 100 g Glass bottle 100g
10787772 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10517021 Supplier Thermo Scientific Acros Supplier No. 349721000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-70-4 | C6H16BrN | 182.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-70-4
Molecular Formula C6H16BrN
Molecular Weight (g/mol) 182.11
MDL Number MFCD00054287
InChI Key NRTLTGGGUQIRRT-UHFFFAOYSA-N
Synonym triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
PubChem CID 2734074
SMILES [Br-].CC[NH+](CC)CC

Specifications

Color White
Flash Point >65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5)3N·HBr
Quantity 100 g
IUPAC Name triethylazanium bromide
Formula Weight 182.1
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Triethylamine hydrobromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Triethylamine hydrobromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.