missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine Hydrochloride, ≥99%, For HPLC, Solstice

Product Code. 15667570 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly Bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15667570 50 g Poly Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15667570 Supplier Honeywell-Fluka Supplier No. 9624950G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
Synonym triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride
PubChem CID 11130
IUPAC Name hydrogen triethylamine chloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point 261°C
Packaging Poly Bottle
Linear Formula (C2H5)3N · HCl
Quantity 50 g
UN Number NONH for all modes of transport
Beilstein 3906409
Formula Weight 137.65g/mol
Percent Purity ≥99%
Chemical Name or Material Triethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.