missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine trihydrofluoride, ca. 37% HF

Product Code. 10081601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081601 25 mL Plastic bottle 25mL
10511194 100 mL Plastic bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081601 Supplier Thermo Scientific Acros Supplier No. 350620250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73602-61-6 | C6H15N·3HF | 161.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73602-61-6
Molecular Formula C6H15N·3HF
Molecular Weight (g/mol) 161.21
MDL Number MFCD00043294
InChI Key IKGLACJFEHSFNN-UHFFFAOYSA-N
Synonym triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
PubChem CID 175505
IUPAC Name N,N-diethylethanamine;trihydrofluoride
SMILES CCN(CC)CC.F.F.F

Specifications

Melting Point -29.0°C
Color Yellow to Brown
Density 0.9890g/mL
Boiling Point 70.0°C (15.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula (C2H5)3N·3HF
Refractive Index 1.3860 to 1.3920
Quantity 25 mL
Specific Gravity 0.989
Solubility Information Solubility in water: soluble
Formula Weight 161.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethylamine trihydrofluoride, ca. 37% HF
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylamine trihydrofluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.