missing translation for 'onlineSavingsMsg'
Learn More

Triethylborane, 1M soln. in THF

Product Code. 11496830
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496830 100 mL 100mL
11486830 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496830 Supplier Thermo Scientific Alfa Aesar Supplier No. L15109.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97-94-9 | C6H15B | 98.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylborane is catalyst for allylation of aldehydes, decarboxylative C-C bond cleavage reactions, rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations, Regioselective hydroxyalkylation of unsaturated oxime ethers. Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential.

Solubility
Reacts with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-94-9
Molecular Formula C6H15B
Molecular Weight (g/mol) 98.00
MDL Number MFCD00009022
InChI Key LALRXNPLTWZJIJ-UHFFFAOYSA-N
Synonym triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
PubChem CID 7357
IUPAC Name triethylborane
SMILES CCB(CC)CC

Specifications

Density 0.865
Flash Point −17°C (1°F)
Linear Formula (CH3CH2)3B
Quantity 100 mL
UN Number UN2924
Beilstein 1731462
Sensitivity Air Sensitive
Solubility Information Reacts with water.
Formula Weight 98
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Triethylborane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethylborane 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, Triethylborane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.