missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Triethylene glycol

Product Code. 16409396
Encompass_Preferred
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16409396 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409396 Supplier MedChemExpress Supplier No. HYW017440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Triethylene glycol (PROTAC Linker 25) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.

Specifications

Molecular Weight (g/mol) 150.17 g/mol
CAS 112-27-6
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 100 mg
Synonym PROTAC Linker 25
Solubility H2O : 100 mg/mL (665.91 mM; Need ultrasonic)
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.