missing translation for 'onlineSavingsMsg'
Learn More

Triethylgermanium chloride, Thermo Scientific™

Product Code. 11387788
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387788 5 g 5g
11377788 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387788 Supplier Thermo Scientific Alfa Aesar Supplier No. 034128.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-28-5
Molecular Formula C6H17ClGe
Molecular Weight (g/mol) 197.28
MDL Number MFCD00013591
InChI Key FJAWDWVPBHGANJ-UHFFFAOYSA-N
Synonym triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane
PubChem CID 70438
IUPAC Name chloro(triethyl)germane
SMILES Cl.CC[GeH](CC)CC

Specifications

Melting Point <-50°C
Density 1.18 g/mL
Boiling Point 175.9°C
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 195.23
Physical Form Liquid
Chemical Name or Material Triethylgermanium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.