missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium Tetrafluoroborate(15% in Dichloromethane, ca. 1mol/L), TRC

Product Code. 30451380
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451380 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451380 Supplier Toronto Research Chemicals Supplier No. TRCT77690025ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyloxonium Tetrafluoroborate
CAS 368-39-8
Molecular Formula C6 H15 O . B F4
Quantity 25 mL
InChI Formula InChI=1S/C6H15O.BF4/c1-4-7(5-2)6-3;2-1(3,4)5/h4-6H2,1-3H3;/q+1;-1
SMILES CC[O+](CC)CC.F[B-](F)(F)F
IUPAC Name triethyloxidanium;tetrafluoroborate
Molecular Weight (g/mol) 189.99
Formula Weight 190.1152
Concentration or Composition (by Analyte or Components) 15 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.