missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, AcroSeal™

Product Code. 10646460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646460 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10646460 Supplier Thermo Scientific Acros Supplier No. 429161000
  • ONLINE EXCLUSIVE PRICE
    135.00 €
    131.00 € / 100mL
    Save 4.00 €
    2% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkyl transfer reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triethyloxonium tetrafluoroborate
CAS 368-39-8, 75-09-2
Color Colorless to Brown
Density 1.3280g/mL
Packaging AcroSeal™ Glass bottle
Molecular Formula C6H15BF4O
Linear Formula (C2H5)3OBF4
MDL Number MFCD00044423
Quantity 100 mL
Merck Index 15, 5867
Specific Gravity 1.328
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
Solubility Information Solubility in water: reacts
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CC[O+](CC)CC
IUPAC Name triethyloxidanium;tetrafluoroborate
Molecular Weight (g/mol) 189.99
PubChem CID 2723982
Formula Weight 189.99
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.