missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 95%

Product Code. 11965921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965921 100 g Glass bottle 100g
11310220 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11965921 Supplier Thermo Scientific Acros Supplier No. 436761000
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkyl transfer reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-39-8 , 60-29-7
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Melting Point 84.0°C to 89.0°C
Color White
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Merck Index 15,5867
Solubility Information Solubility in water: decomposes
Stabilizer 3 to 5 % ether
Formula Weight 189.99
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Triethyloxonium tetrafluoroborate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethyloxonium tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.