missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylsilane, 99%, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
missing translation for 'unitSize'
1 L
100mL
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Boiling Point | 107°C to 108°C |
| Quantity | 100 mL |
| Assay Percent Range | 98.5% min. (GC) |
| Linear Formula | (C2H5)3SiH |
| Flash Point | -3°C |
| Merck Index | 15,9841 |
| Solubility Information | Solubility in water: decomposes |
| Formula Weight | 116.28 |
| Percent Purity | 99% |
| Packaging | AcroSeal™ Glass Bottle |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Triethylsilane
- Danger
- Flammable liquid Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur