missing translation for 'onlineSavingsMsg'
Learn More

(Triethylsilyl)acetylene, 97%

Product Code. 10521253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10521253 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10521253 Supplier Thermo Scientific Acros Supplier No. 368730050
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1777-03-3 | C8H16Si | 140.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1777-03-3
Density 0.7830g/mL
Flash Point 17°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H16Si
Linear Formula (CH3CH2)3SiC≡CH
Refractive Index 1.4310 to 1.4330
MDL Number MFCD00075373
Quantity 5 g
Beilstein 04,IV,393
Specific Gravity 0.783
Synonym triethylsilyl acetylene, ethynyltriethylsilane, triethyl ethynyl silane, triethylsilylacetylene, silane, triethylethynyl, triethylsilyl acetylen, triethylsilyl-acetylene, triethyl-ethynyl-silane
InChI Key FWSPXZXVNVQHIF-UHFFFAOYSA-N
SMILES CC[Si](CC)(CC)C#C
IUPAC Name triethyl(ethynyl)silane
Molecular Weight (g/mol) 140.30
PubChem CID 637947
Formula Weight 140.3
Percent Purity 97%
Chemical Name or Material (Triethylsilyl)acetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.