missing translation for 'onlineSavingsMsg'
Learn More

Trifloxystrobin-d6, TRC

Product Code. 30454873
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454873 10 mg 10mg
30453457 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454873 Supplier Toronto Research Chemicals Supplier No. TRCT77890210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifloxystrobin-d6
CAS 2470226-50-5
Molecular Formula C20H13D6F3N2O4
Quantity 10 mg
Synonym (E,E)-α-(methoxyimino)-2-[[[[1-[3-(trifluoromethyl)phenyl]ethylidene]amino]oxy]methyl]benzeneacetic Acid Methyl Ester-d6, CGA 279202-d6, Compass (strobilurin fungicide)-d6, Consist-d6, Flint (fungicide)-d6, Tega-d6, Trifloxystrobin-d6, Trilex-d6, Twist-d6, Zato-d6
InChI Formula InChI=1S/C20H19F3N2O4/c1-13(14-8-6-9-16(11-14)20(21,22)23)24-29-12-15-7-4-5-10-17(15)18(25-28-3)19(26)27-2/h4-11H,12H2,1-3H3/b24-13+,25-18-/i4D,5D,7D,10D,12D2
SMILES C/C(C1=CC(C(F)(F)F)=CC=C1)=N\OC([2H])([2H])C2=C(/C(C(OC)=O)=N\OC)C([2H])=C([2H])C([2H])=C2[2H]
Molecular Weight (g/mol) 414.41
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.