missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde methyl hemiacetal, 90%, technical

Product Code. 15147432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15147432 100 g Glass bottle 100g
15127432 5 g Glass bottle 5g
15137432 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15147432 Supplier Thermo Scientific Acros Supplier No. 449131000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 431-46-9 | C3H5F3O2 | 130.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-46-9
Molecular Formula C3H5F3O2
Molecular Weight (g/mol) 130.07
MDL Number MFCD00013572
InChI Key GWTBCUWZAVMAQV-UHFFFAOYNA-N
Synonym trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal
PubChem CID 9892
IUPAC Name 2,2,2-trifluoro-1-methoxyethanol
SMILES COC(O)C(F)(F)F

Specifications

Density 1.3600g/mL
Boiling Point 104.0°C to 106.0°C
Flash Point 42°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CF3CH(OCH3)OH
Quantity 100 g
Specific Gravity 1.36
Formula Weight 130.07
Percent Purity 90%
Grade Technical
Chemical Name or Material Trifluoroacetaldehyde methyl hemiacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetaldehyde methyl hemiacetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.