missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a λ, Rat, PE, Clone: A110-1, BD

Brand:  BD Biosciences 557078

Product Code. 10348484

  • ONLINE EXCLUSIVE PRICE
    242.00 €
    232.80 € / Each
    Save 9.20 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The A110-1 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, or rat cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone A110-1 was selected as an isotype control following screening for low background on a variety of mouse and rat tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.

Host Species: Rat
Clone: A110-1
Isotype: IgG2a, <special id='310'/>Immunogen: Trinitrophenol-KLH

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
0.2mg/mL
Rat
Affinity Purified
Undiluted at 4°C
A110-1
PE
Trinitrophenol-KLH
0.1 mg
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.