missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, 99+%, for spectroscopy

Product Code. 11934951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934951 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Artikelnummer. 11934951 Lieferant Thermo Scientific Acros Lieferanten-Nr. 434161000
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    171.00 € / 100mL
    Save 16.00 €
    8% Off
Alle Werbeaktionen anzeigen
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Spezifikation

Absorbance 0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
CAS 76-05-1
Melting Point -15.0°C
Boiling Point 72.0°C
Color Colorless
Density 1.5350 g/mL
Quantity 100 mL
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3
Residue after Evaporation 0.005% max.
Molecular Formula C2HF3O2
Specific Gravity 1.535
Linear Formula CF3CO2H
MDL Number MFCD00004169
Beilstein 02,IV,458
Fieser 10,418; 01,1219; 17,340; 16,83; 15,338; 14,322; 12,529; 11,557; 09,483; 08,503; 07,388; 06,613; 05,695; 04,530; 02,433; 03,305
Merck Index 159847
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether,acetone,ethanol,benzene,,ccl4,hexane,methanol,xylene and hexane
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name 2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 114.02
PubChem CID 6422
ChEBI CHEBI:45892
Formula Weight 114.02
Percent Purity 99+%
Grade Spectroscopy
Chemical Name or Material Trifluoroacetic acid, For Spectroscopy
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Trifluoroacetic acid
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Sicherheitshinweis
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.