missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Absorbance | max. 0.10 at 270 nm, max. 0.9 at 260 nm, max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm |
| CAS | 76-05-1 |
| Melting Point | -15.4°C (literature) |
| Boiling Point | 72°C |
| CAS Max % | 100.0000% |
| Molecular Formula | C2HF3O2 |
| Linear Formula | CF3COOH |
| MDL Number | MFCD00004169 |
| UN Number | UN2699 |
| Beilstein | 742035 |
| Show More |
Safety and Handling
Product Identifier
- Trifluoroacetic acid
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: trifluoroacetic acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?