missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid-d, for NMR, 99 atom % D

Product Code. 10184270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
10mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10184270 25 mL Glass Bottle 25mL
10347870 5 mL Glass Bottle 5mL
10285590 10 mL Glass Bottle 10mL
10311384 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10184270 Supplier Thermo Scientific Supplier No. 166400250
  • ONLINE EXCLUSIVE PRICE
    114.00 €
    104.00 € / 25mL
    Save 10.00 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Color Colorless
Density 1.5000g/mL
Deuterium (D) 98.5% min.
Packaging Glass Bottle
Water 0.1% max. (Coulometric)
Quantity 25 mL
Melting Point -15.0°C
Boiling Point 75.0°C
Linear Formula CF3CO2D
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
Formula Weight 115.03
Grade NMR
Chemical Name or Material Trifluoroacetic acid-d, 99 atom%D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid-d
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.