missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid-d, TRC

Product Code. 30449012
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449012 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449012 Supplier Toronto Research Chemicals Supplier No. TRCT7860371G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifluoroacetic Acid-D
CAS 599-00-8
Purity Grade Notes GC
Molecular Formula C2 D F3 O2
Quantity 1 g
Synonym Acetic acid-d','2,2,2-trifluoro-, Acetic acid-d','trifluoro- (6CI,7CI,8CI,9CI), 2,2,2-Trifluoroacetic acid-d, 2,2,2-Trifluoroacetic acid-d1, Deuteriotrifluoroacetic acid, Deuterotrifluoroacetic acid, Trifluoroacetic acid (CF3COOD), Trifluoroacetic acid-carboxy-d, Trifluoroacetic acid-d, Trifluoroacetic acid-d1, [2H]Trifluoroacetic acid
InChI Formula InChI=1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)/i/hD
SMILES [2H]OC(=O)C(F)(F)F
IUPAC Name deuterio 2,2,2-trifluoroacetate
Molecular Weight (g/mol) 115.0295
Formula Weight 114.9991
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.