missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Eluent additive for LC-MS, Honeywell Fluka™

Product Code. 15655440 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
1 L
Packaging:
Ampoule
Glass bottle
Unit Size:
1 L
10mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655440 1 L Glass bottle 1 L
15618400 10 mL Ampoule 10mL
15665440 5 x 10 mL Ampoule Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15655440 Supplier Honeywell-Fluka Supplier No. 409671L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
IUPAC Name trifluoroacetic acid
Formula Weight 114.02g/mol
Percent Purity ≥99% (GC)
Density 1.489g/mL at 20°C
Quantity 1 L
Packaging Glass bottle
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.