missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™

Product Code. 10224733
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224733 500 mL Amber Glass 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10224733 Supplier Fisher BioReagents Supplier No. BP618500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For peptide synthesis and sequencing protocols

Trifluoroacetic acid is used in peptide sythesis and sequencing protocols.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 0.06 max. at 275nm, 0.03 max. at 300nm
pH 0.45
CAS 76-05-1
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Density 1.535g/cm³
Quantity 500 mL
Packaging Amber Glass
Physical Form Liquid
Assay Percent Range ≥99.5 %
Molecular Formula C2HF3O2
Linear Formula CF3COOH
MDL Number MFCD00004169
Vapor Pressure 107mbar at 25°C
Merck Index 15, 9847
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
PubChem CID 6422
ChEBI CHEBI:45892
Formula Weight 114.02
Percent Purity ≥99.5%
Grade Peptide Synthesis
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.