missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Solstice
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Density | 1.489g/mL at 20°C |
| Quantity | 10 Ampules |
| Flash Point | >100°C (>212°F) |
| Linear Formula | CF3COOH |
| Vapor Density | 3.9 (vs air) |
| UN Number | UN2699 |
| Vapor Pressure | 97.5 mmHg (20°C) |
| Formula Weight | 114.02 |
| Percent Purity | 99% |
| Chemical Name or Material | Trifluoroacetic acid |
Safety and Handling
Product Identifier
- Trifluoroacetic acid
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?