missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Reagent Grade, 99%, Solstice

Product Code. 15688490
Change view
Click to view available options
Quantity:
1 L
10 Ampules
100 mL
2 L
25 mL
5 mL
5 x 10 Ampules
500 mL
Unit Size:
1 L
10 ampoules
100mL
2 L
25mL
500mL
5mL
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15688490 25 mL 25mL
15698490 5 mL 5mL
15668190 100 mL 100mL
15628200 500 mL 500mL
15608200 1 L 1 L
15618200 2 L 2 L
15678190 10 Ampules 10 ampoules
15638200 5 x 10 Ampules Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15688490 Supplier Fluka Supplier No. T650825ML
  • ONLINE EXCLUSIVE PRICE
    46.00 €
    41.18 € / 25mL
    Save 4.82 €
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Density 1.489g/mL at 20°C
Quantity 25 mL
Flash Point >100°C (>212°F)
Linear Formula CF3COOH
Vapor Density 3.9 (vs air)
UN Number UN2699
Vapor Pressure 97.5 mmHg (20°C)
Beilstein 742035
Formula Weight 114.02g/mol
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.