missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, silver salt, 98%

Product Code. 10615002 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10615002

Brand: Thermo Scientific Acros 139730250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2966-50-9 | C2AgF3O2 | 220.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2966-50-9
Molecular Formula C2AgF3O2
Molecular Weight (g/mol) 220.89
MDL Number MFCD00013199
InChI Key KZJPVUDYAMEDRM-UHFFFAOYSA-M
Synonym silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
PubChem CID 76299
IUPAC Name silver;2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].[Ag+]

Specifications

Melting Point 257.0°C to 260.0°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Linear Formula CF3CO2Ag
Quantity 25 g
Fieser 01,1018; 07,323; 08,444; 11,471; 16,301; 17,154
Solubility Information Solubility in water: soluble
Formula Weight 220.89
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetic acid, silver salt, 0.98
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.