missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, Thermo Scientific

Product Code. 30280948 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
5 mL
50 mL
10 x 1 mL
Unit Size:
1mL
2mL
50mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280948 10 x 1 mL 1mL
30280970 1 mL 1mL
30280966 2 mL 2mL
30280947 5 mL 5mL
30280960 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30280948 Supplier Thermo Scientific Supplier No. T0011610X1
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Specifications

Item Name Trifluoroacetic Acid (TFA)
Form Liquid
Reagent Type Ion Pairing Reagent
Quantity 10 x 1 mL
Formulation 99.5% TFA
For Use With LC/MS
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Trifluoroacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.