missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Thermo Scientific™

Product Code. 15517039 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15517039 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15517039 Supplier Thermo Scientific Acros Supplier No. 346610500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 76-05-1
Melting Point -15.0°C
Boiling Point 72.0°C
Molecular Formula C2HF3O2
Linear Formula CF3CO2H
MDL Number MFCD00004169
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
PubChem CID 6422
ChEBI CHEBI:45892
Formula Weight 114.02
Percent Purity 99.5%
Grade Analytical
Density 1.5350g/mL
Quantity 50 mL
Infrared Spectrum Authentic
Refractive Index 1.3
Residue after Evaporation 0.0001% max.
Specific Gravity 1.535
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.