missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic anhydride, 99+%

Product Code. 10498100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
100g
2.5kg
250g
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498100 100 g Glass Bottle 100g
10205150 25 g Glass Bottle 25g
10753337 250 g Glass Bottle 250g
10676342 500 g Glass Bottle 500g
11350290 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10498100 Supplier Thermo Scientific Acros Supplier No. 147811000
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    111.00 € / 100g
    Save 4.00 €
    3% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Trifluoroacetic anhydride, 99+%, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0 | CAS: 407-25-0 | C4F6O3 | 210.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
PubChem CID 9845
IUPAC Name (2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Specifications

Melting Point -63.5°C
Density 1.5100g/mL
Boiling Point 39.1°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CF3CO)2O
Refractive Index 1.3
Quantity 100 g
Beilstein 02, II, 186
Fieser 01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
Formula Weight 210.03
Vapor Pressure 141mbar at 20°C
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Trifluoroacetic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.