missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic anhydride, 98%

Product Code. 11463910
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463910 250 g 250g
11443910 10 g 10g
11439333 25 g 25g
11453910 50 g 50g
11449333 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11463910 Supplier Thermo Scientific Alfa Aesar Supplier No. A11767.30
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

Alfa Aesar Trifluoromethanesulfonic anhydride, 98%, Quantity: 250g, Boiling Point: 81 deg.C to 83 deg.C, Melting Point: -80 deg.C, Molecular Weight: 282.127, Percent Purity: 98%, Beilstein: 1813600, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.72 | CAS: 358-23-6 | C2F6O5S2 | 282.127 g/mol

Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.

Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 358-23-6
Molecular Formula C2F6O5S2
Molecular Weight (g/mol) 282.127
MDL Number MFCD00000408
InChI Key WJKHJLXJJJATHN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
PubChem CID 67749
ChEBI CHEBI:48509
IUPAC Name trifluoromethylsulfonyl trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point -80°C
Density 1.72
Boiling Point 81°C to 83°C
Linear Formula (CF3SO2)2O
Refractive Index 1.321
Quantity 250 g
UN Number UN3265
Beilstein 1813600
Sensitivity Moisture sensitive
Solubility Information Miscible with dichloromethane. Immiscible with hydrocarbons.
Formula Weight 282.13
Percent Purity 98%
Chemical Name or Material Trifluoromethanesulfonic anhydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.