missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF
Shop alle Thermo Scientific Chemicals produkter
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Förpackningsstorlek
100mL
25mL
5mL
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| CAS Min % | 7.15, 91.26 |
| CAS Max % | 8.74, 92.85 |
| Color | Colorless |
| Density | 0.8950g/mL |
| Boiling Point | 40.0°C |
| Flash Point | -20°C |
| Packaging | Glass bottle |
| Linear Formula | (CH3)3SiCF3 |
| Quantity | 5 mL |
| Merck Index | 15, 9851 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- (Trifluoromethyl)trimethylsilane, 0.5M solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion