missing translation for 'onlineSavingsMsg'
Learn More

Trifluorothymidine, 98%

Product Code. 15255073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15255073 250 mg 250mg
15299786 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15255073 Supplier Thermo Scientific Acros Supplier No. 452912500
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 70-00-8 | C10H11F3N2O5 | 296.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-00-8
Molecular Formula C10H11F3N2O5
Molecular Weight (g/mol) 296.20
MDL Number MFCD00006534
InChI Key VSQQQLOSPVPRAZ-RRKCRQDMSA-N
Synonym trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd
PubChem CID 6256
ChEBI CHEBI:75179
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F

Specifications

Melting Point 190°C to 193°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Merck Index 159855
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in isopropanol
Formula Weight 296.2
Percent Purity 98%
Chemical Name or Material Trifluorothymidine
compliance-icons compliance-icons
Product Identifier
  • Trifluorothymidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.