missing translation for 'onlineSavingsMsg'
Learn More

Trifluorothymidine, TRC

Product Code. 30454517
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454517 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454517 Supplier Toronto Research Chemicals Supplier No. TRCT79280050MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifluorothymidine
CAS 70-00-8
Molecular Formula C10 H11 F3 N2 O5
Quantity 50 mg
Synonym Trifluridine, Thymidine','α,α,α-trifluoro-, Uridine','2'-deoxy-5-(trifluoromethyl)- (7CI), α,α,α-Trifluorothymidine, 2'-Deoxy-5-(trifluoromethyl)uridine, 5-(Trifluoromethyl)-2'-deoxyuridine, 5-(Trifluoromethyl)deoxyuridine, 5-Trifluoromethyl-2'-deoxy-β-uridine, 5-Trifluorothymidine, NSC 529182, NSC 75520, Viroptic, 1-(2-Deoxy-β-d-erythro-pentofuranosyl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
InChI Formula InChI=1S/C10H11F3N2O5/c11-10(12,13)4-2-15(9(19)14-8(4)18)7-1-5(17)6(3-16)20-7/h2,5-7,16-17H,1,3H2,(H,14,18,19)/t5-,6+,7+/m0/s1
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C(=O)NC2=O)C(F)(F)F
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)pyrimidine-2,4-dione
Molecular Weight (g/mol) 296.2
Formula Weight 296.062
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.