missing translation for 'onlineSavingsMsg'
Learn More

Trifluralin, MedChemExpress

Product Code. 16402306
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402306 Supplier Medchem Express Supplier No. HYB2050
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Trifluralin is a commonly used pre-emergence herbicide.

Trifluralin is a commonly used pre-emergence herbicide.

Specifications

Chemical Name or Material Trifluralin
CAS 1582-09-8
Color Red
Purity Grade Notes Research
Molecular Formula C13H16F3N3O4
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (298.26 mM) ∣H2O : 0.87 mg/mL (2.59 mM; Need ultrasonic)
SMILES FC(C1=CC([N+]([O-])=O)=C(N(CCC)CCC)C([N+]([O-])=O)=C1)(F)F
Molecular Weight (g/mol) 335.28
Formula Weight 335.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.