missing translation for 'onlineSavingsMsg'
Learn More

Triglyme, TRC

Product Code. 30452298
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452298 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452298 Supplier Toronto Research Chemicals Supplier No. TRCT793800100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triglyme
CAS 112-49-2
Molecular Formula C8 H18 O4
Quantity 100 g
Synonym 2,5,8,11-Tetraoxadodecane, 1,2-Bis(2-methoxyethoxy)ethane, Ansul Ether 161, DMTG, Ethane','1,2-bis(2-methoxyethoxy)-, Glyme 4, Hisolve MTM, Methyltriglyme, NSC 66400, Triethylene glycol dimethyl ether, Triglyme
InChI Formula InChI=1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3
SMILES COCCOCCOCCOC
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
Molecular Weight (g/mol) 178.23
Formula Weight 178.1205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.