missing translation for 'onlineSavingsMsg'
Learn More

Triiron dodecacarbonyl, 99%, stabilized

Product Code. 10115212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10115212 5 g Glass bottle 5g
10416373 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10115212 Supplier Thermo Scientific Acros Supplier No. 209290050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17685-52-8 | C12Fe3O12 | 503.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17685-52-8
Molecular Formula C12Fe3O12
Molecular Weight (g/mol) 503.66
MDL Number MFCD00135617
Synonym triiron dodecarbonyl

Specifications

Melting Point 140.0°C
Color Black
Density 1.9900g/mL
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Fe3(CO)12
Quantity 5 g
Fieser 04,534; 05,706; 07,144; 08,41
Specific Gravity 1.99
Solubility Information Solubility in water: insoluble
Stabilizer 5 to 10% MeOH
Formula Weight 503.66
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Triiron dodecacarbonyl
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triiron dodecacarbonyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H371-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.