Learn More
Triisopropanolamine, 95%, Thermo Scientific Chemicals
Description
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.
Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Melting Point | 48°C to 52°C |
| Boiling Point | 189°C to 191°C (23 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Amine-like |
| Linear Formula | (CH3CHOHCH2)3N |
| Quantity | 10,000 g |
| Beilstein | 1071570 |
| Sensitivity | Hygroscopic |
| Solubility Information | Insoluble in water. Soluble ethanol,diethyl ether,chloroform (slightly ),and methanol (>500g/100g). |
| Formula Weight | 191.27 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.